HardyDisks™ AST Streptomycin, S-10, Hardy Diagnostics
Catalog # 10789-742
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:Streptomycin, S-10, HardyDisks™ AST
- Size:1 x 50 disks
- Cat. No.:10789-742
- Supplier no.:Z8991
Specifications
About this item
Iimpregnated paper disks used for Antimicrobial Susceptibility Testing (AST);
- Spring loaded for smooth and reliable dispensing
- Fully compatible with the BBL single or multiple disk dispensers
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing. The HardyDisk™ AST cartridges are designed to be fully compatible with the BBL single or multiple disk dispensers.