Sabdex (Sabouraud Dextrose) Agar, USP
Catalog # 89407-866
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:PC bottle
- Description:Sabdex (Sabouraud Dextrose) Agar, USP
- Size:200 ml
- Storage Temperature:–2...+30 °C
- Cat. No.:89407-866
- Volume:200 ml
- Supplier no.:U270
Specifications
About this item
For the cultivation of fungi.
(Sabourauds Dextrose Agar) For the cultivation of fungi.