Pyruvate Broth, Hardy Diagnostics
Catalog # 75845-732
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:PC tube
- Description:Pyruvate Broth
- Size:16 x 100 mm
- Cat. No.:75845-732
- Volume:5 ml
- Supplier no.:Y130
Specifications
About this item
For the determination of pyruvate utilization in bacteria.