Order Entry
United States
Orders LinkContactUsLinkComponent
PYR Reagent, Hardy Diagnostics

PYR Reagent, Hardy Diagnostics

PYR Reagent, 15 ml, Dropping vial
Catalog # 75803-062
Supplier:   Hardy Diagnostics
Supplier #:  Z127
PYR Reagent, Hardy Diagnostics
Catalog # 75803-062
Supplier:  Hardy Diagnostics
CAS Number:  
PYR Reagent, Hardy Diagnostics
PYR Reagent, Hardy Diagnostics

PYR Reagent, 15 ml, Dropping vial

CAS Number:  
Price
$48.90
Log in for availability
Each (15ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

For use in conjunction with PYR disks for the detection of pyroglutamate aminopeptidase found in group A streptococci (Streptococcus pyogenes) and group D enterococci.

  • Combines with beta-naphthyl amide
  • Released during pyroglutamate aminopeptidase (PYR) activity to form a pink to red pigment

For use in conjunction with PYR disks for the detection of pyroglutamate aminopeptidase found in group A streptococci (Streptococcus pyogenes) and group D enterococci

The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.

The PYR Reagent combines with beta-naphthyl amide released during pyroglutamate aminopeptidase (PYR) activity to form a pink to red pigment.

Documents

Specifications

Specifications

  • Pack type:
    Dropping vial
  • Description:
    PYR Reagent
  • Volume:
    15 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.