ContactUsLinkComponent
Hygiena PRO-Clean™ Protein Residue Test, Hardy Diagnostics
Hygiena PRO-Clean™ Protein Residue Test, Hardy Diagnostics
  10124-530
Product available on GSA Advantage®
 :  
undefined
Hygiena PRO-Clean™ Protein Residue Test, Hardy Diagnostics
  10124-530
 :  PRO100
Product available on GSA Advantage®
 :  
  • Description:
    Rapid Protein Residue Test, Hygiena, 100/cs
  • Size:
    100 per case
  • Cat. No.:
    10124-530
  • Supplier no.:
    PRO100
A quick and easy way to accurately monitor the cleanliness of food equipment surfaces. Detects protein residues left on a surface after cleaning. Ideal for meat and food processors, restaurants and food service and grocery stores.
  • Results in 1-10 minutes depending on sensitivity required - Instant results allow for immediate corrective action
  • Easy-to-interpret color change
  • All-in-one device- no instrumentation required
  • Quick and easy way to monitor HACCP/SSOP program
A quick and easy way to accurately monitor the cleanliness of food equipment surfaces to help ensure food safety and product quality
Sensitivity: 80 µg after 1 minute, 50 µg after 5 minutes, 20 µg after 10 minutes