Order Entry
United States
ContactUsLinkComponent
Potassium Hydroxide, 10% KOH, Hardy Diagnostics
Potassium Hydroxide, 10% KOH, Hardy Diagnostics
Catalog # 10063-758
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Potassium Hydroxide, 10% KOH, Hardy Diagnostics
Catalog # 10063-758
Supplier:  Hardy Diagnostics
Supplier Number:  Z77
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Dropping vial
  • Description:
    Potassium Hydroxide, 10% KOH
  • Cat. No.:
    10063-758
  • Volume:
    15 ml
  • Supplier no.:
    Z77

Specifications

About this item

For use in slide preparations of specimens for the purpose of identifying fungal elements.

  • Dissolves epithelial cells

For use in slide preparations of specimens for the purpose of identifying fungal elements

The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.

Dissolves epithelial cells that may mask fungal elements.