ContactUsLinkComponent
Copan Plastic square sampling template, Hardy Diagnostics
Copan Plastic square sampling template, Hardy Diagnostics
Catalog # 10024-258
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Copan Plastic square sampling template, Hardy Diagnostics
Catalog # 10024-258
Supplier:  Hardy Diagnostics
Supplier Number:  T2905C
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Description:
    Plastic Square Sampling Template
  • Size:
    10 x 10 cm
  • Cat. No.:
    10024-258
  • Supplier no.:
    T2905C
Specifications
About this item
Plastic square sampling template used for meat carcass testing and/or enumeration procedures.
Plastic square sampling template used for meat carcass testing and/or enumeration procedures. Can be used in conjunction with the Envirotrans™ Environmental Swabing System.