Order Entry
United States
Orders LinkContactUsLinkComponent
 

Phosphate Buffer

Phosphate Buffer, Pack of 10, 500 ml, PC bottle
Catalog # 10124-486
Product available on GSA Advantage®
Supplier:   Hardy Diagnostics
 :  U438
Phosphate Buffer
Catalog # 10124-486
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
Phosphate Buffer
Phosphate Buffer

Phosphate Buffer, Pack of 10, 500 ml, PC bottle

CAS Number:  
Price
$366.00
Log in for availability
Pack of 10 (500ml)
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Recommended for use as a diluent in the preparation of dilutions for plate counts and other laboratory processes for non-clinical samples.

Recommended for use as a diluent in the preparation of dilutions for plate counts and other laboratory processes for non-clinical samples. Not intended for use in the diagnosis of human disease.

Specifications

Specifications

  • Pack type:
    PC bottle
  • Description:
    Phosphate Buffer
  • Size:
    Pack of 10
  • Volume:
    500 ml
  • Cat. No.:
    10124-486
  • pH:
    7.2

Specifications