ContactUsLinkComponent
Phenol Red Broth with Lactose, Hardy Diagnostics
Phenol Red Broth with Lactose, Hardy Diagnostics
  10124-526
Product available on GSA Advantage®
 :  
undefined
Phenol Red Broth with Lactose, Hardy Diagnostics
  10124-526
 :  Y309
Product available on GSA Advantage®
 :  
  • Description:
    Phenol Red Broth with Lactose
  • Size:
    16 x 125 mm
  • Cat. No.:
    10124-526
  • Volume:
    10 ml
  • Supplier no.:
    Y309
For the differentiation of microorganisms by carbohydrate fermentation.
For the differentiation of microorganisms by carbohydrate fermentation. Contains a durham tube for gas detection.