Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Media for Microbiology
Dehydrated Media for Microbiology
Oxoid PALCAM Agar Base, Hardy Diagnostics
Oxoid PALCAM Agar Base, Hardy Diagnostics
Oxoid PALCAM Agar Base, Hardy Diagnostics
Catalog # 10790-404
Supplier:
Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Expand image
Oxoid PALCAM Agar Base, Hardy Diagnostics
Catalog # 10790-404
Supplier:
Hardy Diagnostics
Supplier Number:
CM877B
Product available on GSA Advantage®
CAS Number:
Specifications
Description:
PALCAM Agar Base
Size:
500 gm
Cat. No.:
10790-404
Supplier no.:
CM877B
Specifications
About this item
Product Details
Product Details