Specifications
- Description:Novobiocin, NB-30
- Size:5 x 50 disks
- Environmentally Preferable:
- Cat. No.:10789-456
- Supplier no.:Z8835
Specifications
About this item
Impregnated paper disks used for Antimicrobial Susceptibility Testing (AST)
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing. The HardyDisk™ AST cartridges are designed to be fully compatible with the BBL single or multiple disk dispensers.