Microgen® Bioproducts Microgen® Clostridium Difficile Latex Rapid Test, Hardy Diagnostics
Catalog # 10062-776
Supplier: Avantor
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:Microgen C. Difficile
- Size:50 Tests per Kit
- Cat. No.:10062-776
- Supplier no.:M41
Specifications
About this item
A rapid latex slide agglutination test for the confirmatory identification of Clostridium difficile.
A rapid latex slide agglutination test for the confirmatory identification of Clostridium difficile cultured on selective solid media from fecal samples.
Has a very high specificity and high sensitivity. Use of Microgen® C. difficile latex allows identification of C. difficile from selective agar within two minutes.
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.