Listeria Selective Supplement, Hardy Diagnostics
89423-156
Product available on GSA Advantage®
:
- Description:Listeria Selective Supplement
- Size:10 vials
- Cat. No.:89423-156
- Supplier no.:SR140E
A selective supplement for the isolation of Listeria.
A selective supplement for the isolation of Listeria when used with Listeria Selective Agar Base (Oxford formulation) CM856B. Vial contents (each vial is sufficient for 500ml of medium): Cycloheximide 200mg Colistin sulphate 10mg Acriflavine 2.5mg Cefotetan 1.0mg Fosfomycin 5.0mg