ContactUsLinkComponent
Iodine-Iodide Solution, for Tetrathionate Broth, Hardy Diagnostics
Iodine-Iodide Solution, for Tetrathionate Broth, Hardy Diagnostics
Catalog # 10053-840
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Iodine-Iodide Solution, for Tetrathionate Broth, Hardy Diagnostics
Catalog # 10053-840
Supplier:  Hardy Diagnostics
Supplier Number:  Z139
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Pack type:
    PC bottle
  • Description:
    Iodine-Iodide Solution, HDx
  • Cat. No.:
    10053-840
  • Volume:
    990 ml
  • Supplier no.:
    Z139
Specifications
About this item
For the selective enrichment of Salmonella spp. in clinical and non-clinical specimens.
Iodine-Iodide Solution to be added to Tetrathionate Broth, sold separately
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.
Ordering information: **This is a controlled substance regulated by the DEA, call 800/266-2222