ContactUsLinkComponent
Microgen® Bioproducts E. coli O157 Rapid Latex Slide Agglutination Test, Hardy Diagnostics
Microgen® Bioproducts E. coli O157 Rapid Latex Slide Agglutination Test, Hardy Diagnostics
Catalog # 10062-764
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  
undefined
Microgen® Bioproducts E. coli O157 Rapid Latex Slide Agglutination Test, Hardy Diagnostics
Catalog # 10062-764
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Microgen E.coli O157
  • Size:
    50 Tests
  • Cat. No.:
    10062-764
  • Supplier no.:
    M44
Specifications
About this item
A rapid latex slide agglutination test for identifying E.coli O157 grown on selective medium.
  • Results in two minutes
  • Increases efficiency
  • High sensitivity
  • High specificity
  • Accurate and reliable results
Based on proven and reliable latex agglutination technology, a rapid latex slide agglutination test for identifying E. coli O157 grown on selective medium. Suitable for clinical and food testing.