Order Entry
United States
ContactUsLinkComponent
Pulse Scientific CO₂ Indicator, Hardy Diagnostics
Pulse Scientific CO₂ Indicator, Hardy Diagnostics
Catalog # 10064-880
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
Pulse Scientific CO₂ Indicator, Hardy Diagnostics
Catalog # 10064-880
Supplier:  Hardy Diagnostics
Supplier Number:  G511
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Description:
    CO2 Indicator
  • Size:
    100 per box
  • Cat. No.:
    10064-880
  • Supplier no.:
    G511

Specifications

About this item

Color indicator that detects the atmospheric presence of 5% or more of CO2.

Color indicator that detects the atmospheric presence of 5% or more of CO2.