Oxoid Campylobacter Supplement, Hardy Diagnostics
Catalog # 89423-160
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Vial
- Description:Campy Supplement
- Size:10 Vials
- Cat. No.:89423-160
- Supplier no.:SR69E
Specifications
About this item
A selective supplement for the isolation of campylobacters.
A selective supplement for the isolation of campylobacters when used with Blood Agar Base No.2, Columbia Agar, or Brucella Agar Bases. Vial contents (each vial is sufficient for 500ml of medium): Vancomycin 5mg Trimethoprim 2.5mg Polymyxin B 1250 IU