Butterfield's Phosphate Buffer, Hardy Diagnostics
Catalog # 89407-418
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:PC bottle
- Description:Butterflds Phosphate Buffer
- Size:400 mL
- Storage Temperature:–2-30°C
- Cat. No.:89407-418
- Volume:500 ml
- Supplier no.:U154
Specifications
About this item
For the preparation of dilutions for plate count and other laboratory tests.
For the preparation of dilutions for plate count and other laboratory tests.