Hemostat Blood, Horse, with Heparin, Hardy Diagnostics
Catalog # 10052-794
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
Specifications
- Volume:50 ml
- Description:Blood, Horse w/Heparin
- Cat. No.:10052-794
- Packaging:Bottle, screw cap
- Supplier no.:HB050
Specifications
About this item
Blood, horse, with heparin, screw cap bottle
Our animal blood is obtained from a controlled herd that is kept free of antimicrobic drugs and disease
All types of animal blood and packaging configurations are available.
All animals are maintained in a healthy condition and treated humanely in accordance with government regulations. Fresh blood is available every week.