Medical Action Industries Biohazard Bag, Hardy Diagnostics
10789-388
Product available on GSA Advantage®
- Capacity:2.25 ml
- Description:Biohazard bag, box of 200
- Cat. No.:10789-388
- Supplier no.:BB2305
This 25×35", Biohazard bag is for the collection of biohazard waste.
- Passed the 165 gm dart impact test
- For collection of biohazard waste, not for autoclaving
Made of polyethylene, solid red with black biohazard warnings in English and Spanish, has the international biohazard symbol.
: Meets OSHA requirements.