BCYE Selective Agar with GPCV Antibiotics, Hardy Diagnostics
10142-512
Product available on GSA Advantage®
:
- Pack type:Plate
- Description:BCYE w/GPCV
- Size:15 x 100 mm
- Cat. No.:10142-512
- Volume:26 ml
- Supplier no.:W169
For the selective isolation of Legionella spp.
- Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
- Optically clear polystyrene for distortion free viewing
Buffered Charcoal Yeast Extract Agar with Glycine, Vancomycin, Polymyxin, and Cycloheximide) for the selective isolation of Legionella spp.
Conforms to ISO11371 formulation.