BCYE Selective Agar (Buffered Charcoal Yeast Extract) with CCVC Antibiotics
Catalog # 75845-728
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:BCYE Selective agar with CCVC
- Dimensions:15×100 mm
- Size:10/pack
- Cat. No.:75845-728
- Format:Plate
- Application:Environmental
- Volume:24 ml
- Supplier no.:G170
Specifications
About this item
Buffered Charcoal Yeast Extract (BCYE) selective agar with Cephalothin, Colistin, Vancomycin, and Cycloheximide antibiotics for the selective isolation and cultivation of Legionella from environmental and clinical specimens.