HardyDisks™ AST Bacitracin B-10, Hardy Diagnostics
Supplier: Hardy Diagnostics
Certificates
About this item
Bacitracin B-10 AST for sensitivity tests only, not for Strep ID.
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing. The HardyDisk™ AST cartridges are designed to be fully compatible with the BBL single or multiple disk dispensers.
Frequently Bought Together