ContactUsLinkComponent
AnaeroGRO™ Peptone Yeast Extract Glucose Broth (PYG), Hardy Diagnostics
AnaeroGRO™ Peptone Yeast Extract Glucose Broth (PYG), Hardy Diagnostics
Catalog # 10052-552
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  
undefined
AnaeroGRO™ Peptone Yeast Extract Glucose Broth (PYG), Hardy Diagnostics
Catalog # 10052-552
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Description:
    AnaeroGRO™ Peptone Yeast Extract Glucose Broth (PYG)
  • Size:
    16 x 125 mm
  • Cat. No.:
    10052-552
  • Volume:
    9 ml
  • Supplier no.:
    AG24H
Specifications
About this item
Enriched, non-selective medium recommended for the cultivation and biochemical identification of obligately anaerobic microorganisms.
Hardy Diagnostics AnaeroGRO™ Peptone Yeast Extract Glucose Broth (PYG) is an enriched, non-selective medium recommended for the cultivation and biochemical identification of obligately anaerobic microorganisms. This medium is also suitable for the growth of anaerobic bacteria to be identified by gas liquid chromatography (GLC) analysis as outlined by the CDC.