Order Entry
United States
ContactUsLinkComponent
Anaerobe Nitrate Reagent A, Hardy Diagnostics
Anaerobe Nitrate Reagent A, Hardy Diagnostics
Catalog # 10015-464
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Anaerobe Nitrate Reagent A, Hardy Diagnostics
Catalog # 10015-464
Supplier:  Hardy Diagnostics
Supplier Number:  Z134
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Description:
    Anaerobe Nitrate Rgt A, HDx
  • Size:
    0.5 oz.
  • Cat. No.:
    10015-464
  • Volume:
    15 ml
  • Supplier no.:
    Z134

Specifications

About this item

Anaerobe Nitrate Reagent A, Sulfanilic Acid for nitrate reduction test, 15mL, formulated especially for anaerobic bacteria, ship ground only

Hardy Diagnostics Anaerobe Nitrate Reagents A, B, and C are recommended for use in determining the nitrate reduction reaction of anaerobic bacteria

The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.

This product is to be used with Nitrate Reagent B (Cat. no. Z135)and Nitrate Reagent C (Cat. no. Z73).