ContactUsLinkComponent
Alpet D2 Surface Sanitizer Wipes, Hardy Diagnostics
  10153-332
Product available on GSA Advantage®
undefined
Alpet D2 Surface Sanitizer Wipes, Hardy Diagnostics
  10153-332
 :  SSW0002
Product available on GSA Advantage®
  • Description:
    Alpet D2 Sanitizing Wipes
  • Cat. No.:
    10153-332
  • Packaging:
    Canister (6 x 160 Wipes)
  • Supplier no.:
    SSW0002
Alpet D2 surface sanitizing wipes are a ready-to-use, no-rinse, food-safe, lintless wiping system. Able to clean and sanitize, Alpet D2 surface sanitizing wipes are available in either a 90 count canister (extra heavy duty wipes) or a 160 count canister for less industrial needs. As with all best sanitizers wiping systems, the spring-loaded lid closes on its own to prevent the drying of wipes, helping to eliminate unnecessary waste.
Alpet D2 is effective against Norovirus, Avian influenza H9N2 and is expected to inactivate all influenza A viruses including Pandemic 2009 H1N1 (formerly called swine flu). Treated surfaces must remain wet for 5 minutes.