Order Entry
United States
ContactUsLinkComponent
ALP Arabinose, Modified, Hardy Diagnostics
ALP Arabinose, Modified, Hardy Diagnostics
Catalog # 75803-040
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
ALP Arabinose, Modified, Hardy Diagnostics
Catalog # 75803-040
Supplier:  Hardy Diagnostics
Supplier Number:  Y62
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Tube
  • Description:
    Modified ALP Arabinose, HDx
  • Size:
    13 x 100 mm
  • Minimum Order Quantity:
    5
  • Cat. No.:
    75803-040
  • Volume:
    3 ml
  • Supplier no.:
    Y62

Specifications

About this item

ALP Arabinose for the identification of non-fermentative bacilli.

(Aerobic Low Peptone) For the identification of non-fermentative bacilli. See technical insert.

Ordering information: Minimum Order - 5 packs