Specifications
- Description:Quick-Load Purple Low Molecular Weight DNA Ladder
- Size:125 gel lanes
- Environmentally Preferable:
- Cat. No.:103218-932
- Supplier no.:N0557S
Specifications
About this item
Quick-Load Purple Low Molecular Weight DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing one tracking dye.
- Convenient, ready-to-load format with NEB's proprietary purple loading dye
- Size range: 25 to 766 bp
- Sharp bands
- No UV Shadow
- Easy to identify reference bands
Tracking dye does not leave a shadow under UV light. The DNA Ladder consists of a proprietary plasmid which is digested to completion with appropriate restriction enzymes to yield 11 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The 200 base pair band has increased intensity to serve as a reference point.