- Description:Frosted mico slides
- Cat. No.:75809-508
- Supplier no.:3800380
Micro Slides use high quality sheet glass in a carefully controlled environment, implementing strict specifications and quality control procedures.
- High-quality sheet glass
- Economical
- For general applications
Economical, general purpose slide without painted surface.