ContactUsLinkComponent
Surgipath® FSC 22® Frozen Section Embedding Medium, Leica
Surgipath® FSC 22® Frozen Section Embedding Medium, Leica
  75806-668
 :  Avantor
 :  
undefined
Surgipath® FSC 22® Frozen Section Embedding Medium, Leica
  75806-668
 :  Avantor
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    FSC 22® Frozen Section Embedding Medium
  • Size:
    9 x 118 mL
  • Environmentally Preferable:
  • Cat. No.:
    75806-668
  • Supplier no.:
    3801480
FSC 22® is a water soluble embedding compound used in frozen sectioning.
  • Water soluble
  • For frozen sectioning
  • Bonds and encapsulates specimens
The compound bonds and encapsulates tissue specimens to the object holder for cryosectioning. Available in 9/ 4oz. Squeeze bottles for easy application. It provides excellent sectioning consistency with minimal curling of sections at a working temperature of -20 °C.