- Description:Modified Perls Prussian Blue Iron Stain Kit
- Cat. No.:75810-406
- Supplier no.:38016SS7
Perls’ Prussian Blue Iron Kit stains are used to detect and identify ferric (Fe3+) iron in tissue preparations, blood smears, or bone marrow smears.
- To detect ferric iron
- For use with tissues, blood, or bone marrow
Minute amounts of ferric acid are commonly found in bone marrow and in the spleen.