About this item
Apex Patient Control Box Slides have clearly designated areas for positive control and patient tissues.
- Available in 4 colors
- Positive charged/Hydrophilic adhesion
- Allows control and patient tissue to be on same slide
This hydrophilicity provides a better base for homogeneous covering by aqueous stains and reagents leading to consistent full slide staining.
All slides in the Apex portfolio are excellent for demanding procedures such as standard IHC, Special Stains and HIER techniques. All slides are 1 x 3 x .04”.