


N,N-Dimethylformamide ≥99.8% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:68-12-2
:
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284
:73.09 g/mol
Boiling Pt:152…153 °C (1.013 hPa)
Melting Pt:–61 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:57.5 °C (1.013.25 hPa)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
:GHS02
:GHS08
:GHS07
Specification Test Results
- Assay (GC):min. 99.8 %
- Water (Karl Fischer):max. 0.08 %
- Non-volatile matter:max. 0.001 %
- Suitability for residue analysis:max. 5 ng/l
Specification Test Results
Frequently Bought Together