Order Entry
United States
ContactUsLinkComponent
Amiodarone hydrochloride USP Reference Standard
Health
Amiodarone hydrochloride USP Reference Standard
Catalog # 101094-272
Supplier:  USP
CAS Number:  
Health
Amiodarone hydrochloride USP Reference Standard
Catalog # 101094-272
Supplier:  USP
Supplier Number:  1027302
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    200 mg
  • Cat. No.:
    101094-272
  • Supplier no.:
    1027302

Specifications

About this item

 :
 :681.78 g/mol
UN:3077
ADR:9,III
 :GHS08
CAS Number:19774-82-4
Formula:C₂₅H₂₉I₂NO₃
MDL Number:MFCD00069204