Order Entry
United States
Orders LinkContactUsLinkComponent
Difco™ Cooked Meat Medium, BD

Difco™ Cooked Meat Medium, BD

Difco™ Cooked Meat Medium, 500 g
Catalog # 90001-914
Supplier:   BD
Supplier #:  226730
Difco™ Cooked Meat Medium, BD
Catalog # 90001-914
Supplier:  BD
CAS Number:  
Difco™ Cooked Meat Medium, BD

Difco™ Cooked Meat Medium, 500 g

CAS Number:  
Price
$1,158.30
New Customer Promotional Price
Log in for availability
Each (500g)
Product Details & Documents

Cooked Meat Medium and the enriched medium are used for the cultivation of anaerobes, especially pathogenic clostridia.

  • Muscle protein provides amino acids and other nutrients
  • Muscle provides reducing substances to permit growth of strict anaerobes
  • Growth is indicated by turbidity and presence of gas bubbles

Cooked Meat Medium provides a favorable environment for the growth of anaerobes. The sulfhydryl groups, which exert the reducing effect, are more available in denatured protein; therefore, the meat particles are cooked for use in the medium.

Documents

Specifications

Specifications

  • Description:
    Difco™ Cooked Meat Medium
  • Size:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.