ContactUsLinkComponent
BBL™ Cary-Blair Transport Medium, BD
BBL™ Cary-Blair Transport Medium, BD
  90000-084
 :  BD
 :  
BBL™ Cary-Blair Transport Medium, BD
  90000-084
 :  BD
 :  211102
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    BBL™ Cary-Blair Transport Medium
  • Size:
    500 g
  • Cat. No.:
    90000-084
  • Supplier no.:
    211102
Cary-Blair Transport Medium is a semisolid medium recommended for use in the transportation and preservation of clinical specimens, primarily stool and rectal swabs.
  • Sodium thioglycollate is added to impede oxidation
  • Disodium phosphate acts as a buffering agent
  • Alkaline pH minimizes destruction of bacteria due to acid formation
  • Low nutrient content
This medium possesses a low nutrient content, a low oxidation-reduction potential, and a high pH, making it successful in recovering Salmonella and Shigella.