Difco™ Bile Esculin Agar Base, BD
90005-158
: BD
:
- Description:Difco™ Bile Esculin Agar Base
- Size:500 g
- Cat. No.:90005-158
- Supplier no.:299068
Bile Esculin Agar is used to differentiate enterococci and the Streptococcus bovis gropu from other streptococci.
- Ferric citrate acts as an indicator of esculin hydrolysis
- Oxgall inhibits gram-positive bacteria other than enterococci
Enterococci are able to split esculin, but other streptococci can not, making esculin hydrolysis a valuable procedure in the identification of enterococci.