Specifications
- Description:Difco™ Azide Dextrose Broth
- Size:500 g
- Cat. No.:90003-102
- Supplier no.:238710
Specifications
About this item
Azide Dextrose Broth is used for cultivating streptococci in water and wastewater.
- Beef extract and peptones provide essential nutrients
- Dextrose is a fermentable carbohydrate
- Sodium chloride maintains osmotic equilibrium
- Sodium azide inhibits cytochrome oxidase in gram-negative bacteria
Azide Dextrose Broth is specified for use in the presumptive test of water and wastewater for fecal streptococci by the Multiple-Tube Technique.