About this item
The H57-597 antibody is specific for the beta chain of the mouse T cell Receptor (TCR). This cell surface protein combines with a second protein chain (alpha chain) to form the alpha-beta TCR that is expressed by NK1.1+ thymocytes, NKT cells, and the majority of peripheral T cells. A small number of T cells may express an alternative heteromer of gamma/delta protein chains, known as the g/d TCR. These receptors participate in a complex with CD3, and with the co-receptors CD4 or CD8, to recognize and respond to antigens bound to MHC molecules on antigen-presenting cells. Such interactions promote T cell receptor signaling (T cell activation) and can result in a number of cellular responses including proliferation, differentiation, production of cytokines or activation-induced cell death. The H57-597 antibody is used as a phenotypic marker for T cells expressing the alpha-beta TCR. It is also widely used to cross-link surface TCR and thereby mimic TCR-mediated cell activation or induction of apoptosis. The antibody does not cross-react with cells expressing the g/d TCR.
Type: Primary
Antigen: TCR beta
Clonality: Monoclonal
Clone: H57-597
Conjugation: PE-Cyanine7
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 75834-310
- 75834-308
- Antigen Symbol:
- TCR beta
- TCR beta
- Antigen Name:
- T cell Receptor (TCR) beta chain
- T cell Receptor (TCR) beta chain
- Antigen Synonyms:
- TCRb|TCR-b chain|TCRbeta|TCR-b|b-TCR
- TCRb|TCR-b chain|TCRbeta|TCR-b|b-TCR
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)-Cy7®
- PE (Phycoerythrin)-Cy7®
- Clone:
- H57-597
- H57-597
- Reactivity:
- Mouse
- Mouse
- Host:
- Armenian hamster
- Armenian hamster
- Gene ID:
- 21577
- 21577
- Isotype:
- IgG
- IgG
- Flow Cytometry:
- Yes
- Yes
- Storage Temperature:
- 2-8°C protected from light
- 2-8°C protected from light