Order Entry
United States
ContactUsLinkComponent
Anti-CD3 Mouse Monoclonal Antibody (APC-Cyanine7) [clone: SK7]
Anti-CD3 Mouse Monoclonal Antibody (APC-Cyanine7) [clone: SK7]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD3 Mouse Monoclonal Antibody (APC-Cyanine7) [clone: SK7]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 75834-274
  • 75834-272
  • Antigen Symbol:
  • CD3
  • CD3
  • Antigen Name:
  • CD3 epsilon subunitT cell receptor complex
  • CD3 epsilon subunitT cell receptor complex
  • Antigen Synonyms:
  • Leu-4|T3
  • Leu-4|T3
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)-Cy7®
  • APC (Allophycocyanin)-Cy7®
  • Clone:
  • SK7
  • SK7
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 916
  • 916
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Storage Temperature:
  • 2-8°C protected from light
  • 2-8°C protected from light

Specifications

Related Information

Frequently Bought Together