Order Entry
United States
ContactUsLinkComponent
Anti-CD3e Armenian Hamster Antibody (APC-Cyanine7) [clone: 145-2C11]
Anti-CD3e Armenian Hamster Antibody (APC-Cyanine7) [clone: 145-2C11]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD3e Armenian Hamster Antibody (APC-Cyanine7) [clone: 145-2C11]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 75834-320
  • 75834-322
  • Antigen Symbol:
  • CD3e
  • CD3e
  • Antigen Name:
  • CD3 epsilon subunitT cell receptor complex
  • CD3 epsilon subunitT cell receptor complex
  • Antigen Synonyms:
  • CD3 epsilon
  • CD3 epsilon
  • Antibody Type:
  • Primary
  • Primary
  • Conjugation:
  • APC (Allophycocyanin)-Cy7®
  • APC (Allophycocyanin)-Cy7®
  • Clone:
  • 145-2C11
  • 145-2C11
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Armenian hamster
  • Armenian hamster
  • Gene ID:
  • 12501
  • 12501
  • Isotype:
  • IgG
  • IgG
  • Flow Cytometry:
  • Yes
  • Yes
  • Storage Temperature:
  • 2-8°C protected from light
  • 2-8°C protected from light

Specifications

Related Information

Frequently Bought Together