Specifications
- Description:BBL™ Phenylethyl Alcohol Agar
- Size:500 g
- Cat. No.:90000-222
- Supplier no.:211539
Specifications
About this item
Phenylethyl Alcohol Agar with 5% Sheep Blood is used to inhibit gram-negative bacteria, specifically Proteus species, in specimens containing a mixed bacterial flora.
- Sheep blood enhances the growth of anaerobic bacteria
- Casein digest and soybean meal provide nutrients
- Sodium chloride maintains osmotic equilibrium
- Agar is the solidifying agent
PEA agar is a selective medium that is used for the isolation of gram-positive Staphylococcus and Streptococcus species from clinical specimens or specimens that contain mixtures of bacterial flora.