Order Entry
United States
Orders LinkContactUsLinkComponent
Difco™ Nutrient Agar 1.5%, BD

Difco™ Nutrient Agar 1.5%, BD

Difco™ Nutrient Agar 1.5%, 500 g
Catalog # 90003-812
Supplier:   BD
Supplier #:  269100
Difco™ Nutrient Agar 1.5%, BD
Catalog # 90003-812
Supplier:  BD
CAS Number:  
Difco™ Nutrient Agar 1.5%, BD

Difco™ Nutrient Agar 1.5%, 500 g

CAS Number:  
Price
$510.84
New Customer Promotional Price
Log in for availability
Each (500g)
Minimum order quantity is 1
Product Details & Documents

Nutrient Agar 1.5% is used for cultivating a variety of microorganisms and with the addition of blood or other enrichment can be used for cultivating fastidious microorganisms.

  • Beef extract provides essential nutrients
  • Sodium chloride maintains the osmotic balance
  • Agar is the solidifying agent

Nutrient Agar 1.5% is a slightly alkaline modification of Nutrient Agar and is a general-purpose medium. Since the medium contains 0.8% sodium chloride, it can be used as a base for enrichment with blood, ascitic fluid, or other supplements for cultivating fastidious microorganisms.

Documents

Specifications

Specifications

  • Description:
    Difco™ Nutrient Agar 1.5%
  • Size:
    500 g

Specifications