To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Beta Galactosidase (β-Gal) Activity Assay Kit (Fluorometric)
- Size:100 assays
- Environmentally Preferable:
- Sample Type:Prokaryote such as E.coli; Animal tissues such as spleen etc.; Adherent or suspension cells; Food such as yogurt; White blood cells
- Detection Method:Fluorescence
- Shelf Life:12 Months
- Ex/em:480/520 nm
- Cat. No.:75832-880
- Storage temperature:–20 °C
- Supplier no.:K821-100
Specifications
About this item
The most sensitive assay to measure β-Gal activity.
- Measurement of β-Galactosidase activity in various samples
- Simple, sensitive, and high-throughput adaptable
- Quick and easy way for monitoring Beta Galactosidase activity
Beta Galactosidase (β-Gal, EC: 3.2.1.23) is an enzyme which hydrolyzes the β-galactosides into monosacchanides. β-Gal is widely used as a reporter gene in the field of molecular biology. Senescence Associated β-Gal (SA-β-Gal) is an isoform of β-Gal which has the optimal activity at pH 6.0, and is mostly used as a biomarker for senescent cells (K802). β-Gal is an essential enzyme in humans and its deficiency results in Morquio’s Syndrome, a severe birth defect. BioVision’s Beta Galactosidase Activity Assay kit provides a quick and easy way for monitoring Beta Galactosidase activity in a variety of samples. In this kit, Beta Galactosidase hydrolyses a non-fluorescent substrate to generate a strong fluorescent product. The assay is simple, sensitive, and high-throughput adaptable. Detection limit: < 0.1 mU.