Order Entry
United States
ContactUsLinkComponent
PicoProbe™ Dihydroxyacetone Phosphate (DHAP) Fluorometric Assay Kit, BioVision
PicoProbe™ Dihydroxyacetone Phosphate (DHAP) Fluorometric Assay Kit, BioVision
Catalog # 75832-910
Supplier:  Avantor
CAS Number:  
undefined
PicoProbe™ Dihydroxyacetone Phosphate (DHAP) Fluorometric Assay Kit, BioVision
Catalog # 75832-910
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    PicoProbeTM Dihydroxyacetone Phosphate (DHAP) Fluorometric Assay Kit
  • Size:
    100 assays
  • Environmentally Preferable:
  • Sample Type:
    Animal tissues: e.g., liver, muscle etc.

    Cell culture: adherent or suspension cells

    Biological fluids: serum, plasma etc.
  • Detection Method:
    Fluorescence
  • Shelf Life:
    12 Months
  • Ex/em:
    535/587 nm
  • Cat. No.:
    75832-910
  • Storage temperature:
    –20 °C
  • Supplier no.:
    K673-100

Specifications

About this item

Measurement of Dhydroxyacetone Phosphate in various tissues/cells.

  • Measurement of Dhydroxyacetone Phosphate in various tissues/cells
  • Analysis of metabolic pathways such as glycolysis, Calvin cycle in plants, methylglyoxal pathway and lipid biosynthesis
  • Mechanistic study of human TPI deficiency

Dihydroxyacetone Phosphate (DHAP) is an important intermediate in both lipid biosynthesis and glycolysis. In glycolysis, Fructose-1,6-diphosphate is converted to DHAP and glyceraldehyde-3-phosphate (GAP) by aldolase. Both DHAP and GAP serve as the intracellular triose phosphate pool. DHAP can be further converted into GAP by Triose Phosphate Isomerase (TPI). In humans, TPI deficiency is a rare autosomal disease. It causes hemolytic anemia, neurological diseases, and even death due to blockage of the glycolytic pathway and accumulation of DHAP in erythrocytes. Biovision’s PicoProbeTM Dihydroxyacetone Phosphate Assay kit is suitable for measuring low levels of DHAP typically found in a variety of samples. In this kit, TPI converts DHAP to GAP that undergoes a series of reaction and reduces PicoProbe™ to generate fluorescence. The fluorescence intensity generated is directly proportional to the amount of Dihydroxyacetone Phosphate. This simple, sensitive, and easy to use assay kit can detect Dihydroxyacetone Phosphate as low as 0.5 µM in a variety of samples.