ContactUsLinkComponent
Thiol resin (MP), beads, macroporous 330-1225 µm, 15-50 mesh Loading: 4.8-5.0 mmol/g
Thiol resin (MP), beads, macroporous 330-1225 µm, 15-50 mesh Loading: 4.8-5.0 mmol/g
Supplier:  Supra Sciences
Certificates
undefined
Thiol resin (MP), beads, macroporous 330-1225 µm, 15-50 mesh Loading: 4.8-5.0 mmol/g
Supplier:  Supra Sciences
Specification Test Results
  • Colour:
    Off-white
  • Form:
    Granule
  • Loading:
    4.8-5.0 mmol/g
  • Particle size:
    330-1225 μm (15-50 mesh)