Sulfonic acid polystyrene resin (MP), beads, macroporous 400-1100 µm, 20-40 mesh(>90% within) Loading: 4.8-5.2 mmol/g
:
:MFCD02683442
- Colour:Off-white
- Form:Granule
- Loading:4.8-5.0 mmol/g
- Particle size:400-1100 μm (20-24 mesh)
Frequently Bought Together