Piperazine on resin (MP), beads, macroporous 100-200 mesh Loading: 1-1.2mmol/g
Supplier: Supra Sciences
Certificates
About this item
CAS Number:110-85-0
:
Formula:C₄H₁₀N₂
MDL Number:MFCD00005953
:86.14 g/mol
Specification Test Results
- Colour:Off-white
- Form:Granule
- Loading:1.0-1.2 mmol/g
- Particle size:100-200 mesh
Specification Test Results
Frequently Bought Together