ContactUsLinkComponent
Carbonate resin (MP), beads, macroporous 330-1225 µm, 15-50 mesh (>90% within) Loading: 2.5-2.8 mmol/g
undefined
Carbonate resin (MP), beads, macroporous 330-1225 µm, 15-50 mesh (>90% within) Loading: 2.5-2.8 mmol/g
  • Colour:
    Off-white
  • Form:
    Granule
  • Loading:
    2.5-2.8 mmol/g
  • Particle size:
    330-1225 μm (15-50 mesh)
Frequently Bought Together