Aminomethyl resin on polystyrene (PS), beads 100-200 mesh Loading: 1.0-1.2 mmol/g
Supplier: Supra Sciences
Certificates
About this item
CAS Number:78578-28-6
:
MDL Number:MFCD00130502
Specification Test Results
- Colour:Off-white
- Form:Granule
- Loading:0.9-1.0 mmol/g
- Particle size:100-200 mesh
Specification Test Results
Frequently Bought Together